Learn More
Eppendorf™ Temperature Sensor Device for Mastercycler
Rapid, easy to use, and suitable for use with Mastercycler PCR cyclers.
Supplier:  Eppendorf™ 0056001000
 View more versions of this product
Catalog No. 05-414-218
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • Measuring device without temperature sensor
  • Sensor for 96 well and 384 well blocks, sold separately
  • Simple, rapid verification and adjustment
  • Automatic adjustment with Mastercycler software, version 2.10 or higher
  • Can be third-party adjusted
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Requires Temperature Sensor
Mastercycler, 96-well
Single Channel
Product Suggestions
Product Suggestions